Ribonuclease A
Glass bottle, 5 mL
Supplier: OMEGA BIO-TEK
Jump to:
Product Details & Documents
- Enzyme activity is 100 mg/ml (for 5 ml)
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:5 mL
- Cat. No.:PD090
- Storage conditions:Freezer
- Enzyme synonyms:RNase A
- MDL No.:MFCD00132184
- Enzyme name:Ribonuclease A
- CAS No.:9001-99-4
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



