0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Rainin SR-L300F Tips Filter 20-300µLStableRak pk768 LTS 1 * 768 items
Catalog # RAINSR-L300F
Supplier:
RAININ
Expand image
Rainin SR-L300F Tips Filter 20-300µLStableRak pk768 LTS 1 * 768 items
Catalog # RAINSR-L300F
Supplier:
RAININ