Methyl violet 2B
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 8004-87-3 Molecular Formula: C24H27N3.HCl Formula Weight: 393.96 Odour: Odourless MDL No.: MFCD00012568
Synonyms: Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
- CAS Number:8004-87-3
- Formula:C₂₄H₂₈ClN₃
- MDL Number:MFCD00012568
- Molecular Weight:393.96 g/mol
- Melting Pt:137 °C
- Storage Temperature:Ambient
- UN:3077
- ADR:9,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



