Order Entry
Sweden
Orders LinkContactUsLinkComponent
Cuvettes and tubes for Picco, Multy, Nova and Prove
Cuvettes and tubes for Picco, Multy, Nova and Prove
Rectangular cuvettes, 10 mm, Quartz
Catalog # 1.00784.0001
Supplier:   Merck
Cuvettes and tubes for Picco, Multy, Nova and Prove
Catalog # 1.00784.0001
Supplier:  Merck
Cuvettes and tubes for Picco, Multy, Nova and Prove
Cuvettes and tubes for Picco, Multy, Nova and Prove
Rectangular cuvettes, 10 mm, Quartz
Price
5 030,00 kr
Log in for availability
1 * 2 Items
Product Details & Documents
Specifications
  • Path length:
    10 mm
  • Window material:
    Quartz
  • Description:
    Rectangular cuvettes
  • Seq:
    04
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • TOC, screw caps for digestion
    TOC, screw caps for digestion
    1.73500.0001
    Log in for availability
    1 * 6 Items - 1 520,00 kr
Product Information
  • Catalog #
    Availability
    Price
  • Arsenic absorption tube with NS 29 joint
    Arsenic absorption tube with NS 29 joint
    1.73501.0001
    Log in for availability
    1 * 1 Items - 889,00 kr