NATRIUMCITRATBUFFER PH 3,2 1 * 2 l
Catalog # WAPR80-2037-68
Supplier: Biochrom
Frequently Bought Together
VWR® pHenomenal® CO 3100 H Conductivity/TDS/°C Meter, Handheld
Supplier: VWR Collection
Accessories for spiral plater/diluter, easySpiral®
Supplier: INTERSCIENCE
VWR® pH CO 1030, pH/Conductivity Tester
Supplier: VWR Collection
VWR®, Hand Tally Counter
Supplier: VWR Collection
VWR®, Halogen Bulbs
Supplier: VWR Collection
Single gas detectors, BW Clip
Supplier: BW TECHNOLOGIES
VWR® pHenomenal® pH 1100 H pH/mV/°C Meter, Handheld
Supplier: VWR Collection
Accessories for flow cups with interchangeable nozzle
Supplier: Industrial Physics
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Accessories for Mettler balances
Supplier: Mettler - Toledo
NATRIUMHYDROXID OPL#SNING 1 * 1 l
Supplier: Biochrom
Lithium Buffer 1 1 * 2 L
Supplier: Biochrom
Sodium Hydro/Oxid Buffer 2 1 * 2 L
Supplier: Biochrom
Plastic cuvette for LICO
Supplier: Hach
Sodium Hydrolysate Buffer 5 1 * 2 L
Supplier: Biochrom