0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Acculab Atilon Series balance, verified version 80G X 0.0001G with internal calibration weight, includes RS232 interface 1 * 1 items
Catalog # SARBATL-84-V
Supplier:
Sartorius Balances
Expand image
Acculab Atilon Series balance, verified version 80G X 0.0001G with internal calibration weight, includes RS232 interface 1 * 1 items
Catalog # SARBATL-84-V
Supplier:
Sartorius Balances