Order Entry
Sweden
ContactUsLinkComponent
Compound microscope, Monocular 10x 3 Obj 4/10/40x Mech stage Abbe Iris LED with battery. Swiss plug. 1 * 1 items
Catalog # 630-0783
Supplier:  VWR Collection
Compound microscope, Monocular 10x 3 Obj 4/10/40x Mech stage Abbe Iris LED with battery. Swiss plug. 1 * 1 items
Catalog # 630-0783
Supplier:  VWR Collection