Order Entry
Sweden
ContactUsLinkComponent
Refrigerator cbp4056 total capacity 406l fridge 315l freezer 91l height 1982 x width 600 x depth 632 mm biofresh 1 * 1 items
Catalog # 471-0167
Leverantör:  Liebherr
Refrigerator cbp4056 total capacity 406l fridge 315l freezer 91l height 1982 x width 600 x depth 632 mm biofresh 1 * 1 items
Catalog # 471-0167
Leverantör:  Liebherr