Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Double-D Autostill waterdistillation unit 4L/hr, 240V 1 * 1 items
Catalog # 171-0247
Supplier:
JENCONS SCIENTIFIC
Expand image
Double-D Autostill waterdistillation unit 4L/hr, 240V 1 * 1 items
Catalog # 171-0247
Supplier:
JENCONS SCIENTIFIC