Overalls, Proshield® 8 Proper, model CCF5
Grey, L
Catalog # DUPOCCF5SGY00/L
Supplier: DuPont
Jump to:
Product Details & Documents
ProShield® Proper overalls are made of a proprietary DuPont non woven PE fabric (55 g/m²) that makes them incredibly tough, lightweight and resistant to tears and punctures.
- Mandarin collar, nylon zipper with flap
- Two thigh pockets
- Stitched seams
- Wrist and back elastication for good fit, open ankles (not elasticated)
- Antistatically treated for comfort (not acc. EN 1149-5)
- Can be re-used several times and may also be washed up to seven times
The overalls are comfortable because the material allows the passage of air and water vapour. They are ideal workwear for do-it-yourself, general maintenance and cleaning, manufacturing and other non hazardous applications as well as a semi re-usable garment for visitors.
Colour: Grey
: Cat. I
Specifications
- Clothing Size:L
- Colour:Grey
- Cat. No.:DUPOCCF5SGY00/L
- Model:CCF5
- Anti-static:+
- Garment application:General Purpose|Food
- Weight:55 g/m²