Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Inlet liners

Splitless, gooseneck, with wool, for trace samples >2 µl, 6,5 mm, 4,0 mm, 78,5 mm
  568-2366
Inlet liners
  568-2366
 :  
Inlet liners
Inlet liners

Splitless, gooseneck, with wool, for trace samples >2 µl, 6,5 mm, 4,0 mm, 78,5 mm

 

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

 
  • Ext.Ø:
    6,5 mm
  • Ø int.:
    4,0 mm
  • Description:
    Splitless, gooseneck, with wool, for trace samples >2 µl
  • Length:
    78,5 mm
  • Seq:
    013