Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Inlet liners

Drilled uniliner for trace, active samples, high recovery and linearity, 6,3 mm, 4,0 mm, 78,5 mm
Katalog # REST20756
Inlet liners
Katalog # REST20756
Leverantör:  
Inlet liners
Inlet liners

Drilled uniliner for trace, active samples, high recovery and linearity, 6,3 mm, 4,0 mm, 78,5 mm

Discontinued
Produktdetaljer och dokument

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    6,3 mm
  • Ø int.:
    4,0 mm
  • Description:
    Drilled uniliner for trace, active samples, high recovery and linearity
  • Length:
    78,5 mm
  • Seq:
    023
  • Cat. No.:
    REST20756