Order Entry
Sweden
Orders LinkContactUsLinkComponent
Experiment kit, electric field lines
Experiment kit, electric field lines
Two bullets
Catalog # 765-1196
Marketsource
Supplier:   PHYWE
Experiment kit, electric field lines
Catalog # 765-1196
Supplier:  PHYWE
Marketsource
Experiment kit, electric field lines
Experiment kit, electric field lines
Two bullets
Price
705,63 kr
Log in for availability
1 * 1 Items
Product Details & Documents
For demonstration of electric fields between electrodes of different shapes. The electrodes have been applied in thin conductive layers on plexiglass plates and the field line images appear in groats placed in a cuvette with 1 to 2 mm castor oil (the cuvette is placed on the plexiglass plate). The cuvette has an annular electrode in the outer jacket and a 4 mm connection sleeve. The cleaning is easy when electrodes resp. plexiglass plate never comes into contact with the oil. Suitable voltage source: High voltage generator 0 to 25 kV or Van de Graff generator.
  • Dimensions: plates 180 × 180 mm, diam cuvette 120 mm
Specifications
  • Description:
    Two bullets