Inlet liners
Cup splitter for high and low MW compounds, 6,3 mm, 4,0 mm, 78,5 mm
Catalog # REST20709
Jump to:
Discontinued
Product Details & Documents
These liners can be used with Agilent GCs.
Ø int. = Nominal diameter at syringe needle expulsion point
Specifications
- Ext.Ø:6,3 mm
- Ø int.:4,0 mm
- Description:Cup splitter for high and low MW compounds
- Length:78,5 mm
- Seq:021
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



