Inlet liners
Laminar cup splitter for high MW compounds, 6,3 mm, 4,0 mm, 78,5 mm
Catalog # REST20801
Jump to:
Discontinued
Product Details & Documents
These liners can be used with Agilent GCs.
Ø int. = Nominal diameter at syringe needle expulsion point
Specifications
- Ext.Ø:6,3 mm
- Ø int.:4,0 mm
- Description:Laminar cup splitter for high MW compounds
- Length:78,5 mm
- Seq:031
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



