Order Entry
Sweden
Orders LinkContactUsLinkComponent
Inlet liners

Inlet liners

Cyclo double taper, for trace, active and dirty samples >2 µl, 6,5 mm, 4,0 mm, 78,5 mm
Catalog # REST20895
Inlet liners
Catalog # REST20895
Supplier:  
Inlet liners
Inlet liners

Cyclo double taper, for trace, active and dirty samples >2 µl, 6,5 mm, 4,0 mm, 78,5 mm

Discontinued
Product Details & Documents

These liners can be used with Agilent GCs.

Ø int. = Nominal diameter at syringe needle expulsion point

Specifications
  • Ext.Ø:
    6,5 mm
  • Ø int.:
    4,0 mm
  • Description:
    Cyclo double taper, for trace, active and dirty samples >2 µl
  • Length:
    78,5 mm
  • Seq:
    033
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.