Order Entry
Sweden
Orders LinkContactUsLinkComponent
Inserts for trays
Inserts for trays
Back insert, 8 compartments Inside dimensions: 120 × 70 × 35 mm
Catalog # 765-0961
Supplier:   GRATNELL'S
Inserts for trays
Catalog # 765-0961
Supplier:  GRATNELL'S
Inserts for trays
Inserts for trays
Back insert, 8 compartments Inside dimensions: 120 × 70 × 35 mm
Price
325,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Inserts made from light grey polystyrene with 3, 4, 6 or 8 sections. Suitable for use with trays F/1, F/2 and R/3.
Specifications
  • Description:
    Back insert, 8 compartments Inside dimensions: 120 × 70 × 35 mm