Order Entry
Sweden
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

Trays, Green, 312×427×75 mm
Catalog # 763-0301
Supplier:   GRATNELL'S
Storage Boxes
Catalog # 763-0301
Supplier:  GRATNELL'S
Storage Boxes
Storage Boxes

Trays, Green, 312×427×75 mm

Price
325,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Polypropylene storage trays. The material is fire-resistant and resistant to most chemicals in the school environment. Stackable.

  • BSI tested and passed for heavy educational use
  • All trays fit standard school furniture
  • Lids and inserts also available
  • Fully recyclable
Specifications
  • Colour:
    Green
  • Description:
    Trays
  • W×D×H:
    312×427×75 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.