Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TUBING, PEEK, 1/32'' OD X 0.25 MM ID, NA 1 * 3 m
Catalog # JOURJR-T-5995-M3
Supplier:
VICI JOUR RESEARCH
Expand image
TUBING, PEEK, 1/32'' OD X 0.25 MM ID, NA 1 * 3 m
Catalog # JOURJR-T-5995-M3
Supplier:
VICI JOUR RESEARCH