ChromBox kit
10-32 nuts, 19 mm, stainless steel
Catalog # JOURJR-LZN1-5
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
The ChromBox is an all-purpose analytical spare parts kit for HPLC. It contains the most often used spare parts made both of high quality Type 316L SS as well as PEEK and PTFE.
: Kit includes PEEK, PTFE, and stainless steel tubing, PEEK and stainless steel nuts, ferrules and unions, tubing cutters and wrenches, all in a convenient carrying case.
Specifications
- Description:10-32 nuts, 19 mm, stainless steel
- Seq:09
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



