Order Entry
Sweden
Orders LinkContactUsLinkComponent
Protein G
Protein G
Protein G, 500 µG
Catalog # ROCKPG00-00
Protein G
Catalog # ROCKPG00-00
CAS Number:  
Protein G
Protein G
Protein G, 500 µG
CAS Number:  
Price
7 550,00 kr
Log in for availability
1 * 500 µG
Product Details & Documents
Protein G is a surface protein of two groups of streptococcal bacteria that has ability to bind immunoglobulins and also to albumins. Protein G are prepared from chromatographically pure protein G.
Protein G is stable at 4 °C as an undiluted liquid. Freezing alkaline phosphatase conjugates will result in a substantial loss of enzymatic activity.
Protein G alkalline phosphatase conjugated is ideal for signal transduction, molecular biology, cancer, and cell signaling. Protein G peroxidase conjugated is a useful reagent in western blotting and ELISA experiments. Protein G fluorescein conjugated is suitable for immunomicroscopy and flow cytometry or FACS analysis.
Documents
Specifications
Specifications
  • Pk:
    500 µG
  • Description:
    Protein G