0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Paar DMA38 Density Meter, +/- 0.001G/cm3 Accuracy, Range 0-3 g/cm3. 1 * 1 items
Catalog # PAARDMA38
Supplier:
Anton Paar
Expand image
Paar DMA38 Density Meter, +/- 0.001G/cm3 Accuracy, Range 0-3 g/cm3. 1 * 1 items
Catalog # PAARDMA38
Supplier:
Anton Paar