Order Entry
Sweden
ContactUsLinkComponent
Thermometer, mercury, BS1704 F/75, white backed, measuring range:0:250/1C, length: 300mm, 75mm immersion, durable pigment, suitabl e for government verification. Until 10th October 2017 it is still allowed to use mercury thermometers if they are ex 1 * 1 items
Catalog # 620-0046
Supplier:  Amarell
Thermometer, mercury, BS1704 F/75, white backed, measuring range:0:250/1C, length: 300mm, 75mm immersion, durable pigment, suitabl e for government verification. Until 10th October 2017 it is still allowed to use mercury thermometers if they are ex 1 * 1 items
Catalog # 620-0046
Supplier:  Amarell