0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Scissors,spring bow handles,stainless steel,120mm 1 * 1 items
Catalog # 233-0019
Leverantör:
MURRAY S
Expand image
Scissors,spring bow handles,stainless steel,120mm 1 * 1 items
Catalog # 233-0019
Leverantör:
MURRAY S