2-(Ethylthio)nicotinamide
Glass bottle for solids, 10 g
Catalog # MAYBBTB06390EA
Marketsource
Supplier: Thermo Scientific
2-(Ethylthio)nicotinamide
Catalog # MAYBBTB06390EA
Supplier: Thermo Scientific
Marketsource
CAS Number: 175135-27-0
Jump to:
Product Details & Documents
- CAS Number:175135-27-0
- Formula:C₈H₁₀N₂OS
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



