Order Entry
Sweden
ContactUsLinkComponent
Thermometer, Anschutz milk glass type, mercury filled, DIN 12777, measuring range:148:202/0.2C, enclosed scale, capillary lens fro n t, colourless, dimension: 175x5-6mm, total immersion, Richter-top finish, suitable for government verification. Unt 1 * 1 items
Catalog # AMARN60166
Supplier:  Amarell
Thermometer, Anschutz milk glass type, mercury filled, DIN 12777, measuring range:148:202/0.2C, enclosed scale, capillary lens fro n t, colourless, dimension: 175x5-6mm, total immersion, Richter-top finish, suitable for government verification. Unt 1 * 1 items
Catalog # AMARN60166
Supplier:  Amarell