Order Entry
Sweden
Orders LinkContactUsLinkComponent
VWR® SuperFrost® Plus, Adhesion Slides

VWR® SuperFrost® Plus, Adhesion Slides

White, 1 mm, 75×25 mm, 75×25 mm
Catalog # 631-0108
Supplier:   VWR Collection
VWR® SuperFrost® Plus, Adhesion Slides
Catalog # 631-0108
Supplier:  VWR Collection
VWR® SuperFrost® Plus, Adhesion Slides
VWR® SuperFrost® Plus, Adhesion Slides

White, 1 mm, 75×25 mm, 75×25 mm

Price
330,00 kr
Log in for availability
1 * 72 Items
Product Details & Documents

Superfrost Plus slides are specially treated for frozen tissue sections.

  • Permanent positive charge binds freshly frozen tissue sections and cytology preparations
  • Ground edges and colour-frosted ends for labelling
  • Tissue section thickness: 2 to 5 µm

Electrostatically binds tissue sections to the slide virtually preventing any loss during staining.

Cytology preparations placed on these slides resist cell loss during enzyme digestion, DNA denaturation, and RNA hybridisation.

Specifications
  • W×D:
    75×25 mm
  • Colour:
    White
  • L×W:
    75×25 mm
  • Seq:
    01
  • Thickness:
    1 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.