Order Entry
Sweden
ContactUsLinkComponent
Thermometer, industrial, straight stem, mercury filled, measuring range:0:100/1C, capillary lens front, yellow backed, milk glass s cale, upper part:235x20mm, lower part: 255x8mm, cork-glass-top finish. Until 10th October 2017 it is still allowed t 1 * 1 items
Catalog # AMART90016
Supplier:  Amarell
Thermometer, industrial, straight stem, mercury filled, measuring range:0:100/1C, capillary lens front, yellow backed, milk glass s cale, upper part:235x20mm, lower part: 255x8mm, cork-glass-top finish. Until 10th October 2017 it is still allowed t 1 * 1 items
Catalog # AMART90016
Supplier:  Amarell