Order Entry
Sweden
Orders LinkContactUsLinkComponent
ChromBox kit
ChromBox kit
Tweezers, PP
Catalog # JOURJR-806
Supplier:   VICI JOUR RESEARCH
ChromBox kit
Catalog # JOURJR-806
Supplier:  VICI JOUR RESEARCH
ChromBox kit
Tweezers, PP
Price
235,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
The ChromBox is an all-purpose analytical spare parts kit for HPLC. It contains the most often used spare parts made both of high quality Type 316L SS as well as PEEK and PTFE.
Ordering information: Kit includes PEEK, PTFE, and stainless steel tubing, PEEK and stainless steel nuts, ferrules and unions, tubing cutters and wrenches, all in a convenient carrying case.
Specifications
  • Description:
    Tweezers, PP
  • Seq:
    06