Order Entry
Sweden
Orders LinkContactUsLinkComponent
Luminescent bacteria test kit

Luminescent bacteria test kit

NaCl solution 2% for luminescent bacteria test, 250 mL
Catalog # HACHLCK481
Supplier:   Hach
Luminescent bacteria test kit
Catalog # HACHLCK481
Supplier:  Hach
CAS Number:  
Luminescent bacteria test kit
Luminescent bacteria test kit

NaCl solution 2% for luminescent bacteria test, 250 mL

CAS Number:  
Price
555,00 kr
Log in for availability
1 * 250 mL
Product Details & Documents

The luminescent bacteria test is a simple operation to carry out as a chemical analysis. It provides quick results within maximum test time of 30 min.

  • Long-life preserved bacteria
  • Viability indicated by natural luminescence
  • Effortless compensation for interferences

The preserved luminescent bacteria are reactivated before the test is started. Toxicity is indicated by the luminescence, more the sample inhibits luminescence, more toxic it is.

Certifications: Conforms to international EN ISO 11348 standard.

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    NaCl solution 2% for luminescent bacteria test

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.