0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
LiChroCART 250-10 Purospher STAR RP-18e 5µm 1 * 1 Each
Catalog # 1.50179.7185
Supplier:
Merck
SDS
Expand image
LiChroCART 250-10 Purospher STAR RP-18e 5µm 1 * 1 Each
Catalog # 1.50179.7185
Supplier:
Merck
SDS
Related Information
Documentation
SDS
msds_150179
Documentation