Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Dropping bottles, LDPE, WHEATON®

60 ml, LDPE, white, 20-410
Catalog # 215-4003
Supplier:   DWK Life Sciences
Dropping bottles, LDPE, WHEATON®
Catalog # 215-4003
Supplier:  DWK Life Sciences
Dropping bottles, LDPE, WHEATON®
Dropping bottles, LDPE, WHEATON®

60 ml, LDPE, white, 20-410

Price
6 725,00 kr
Log in for availability
1 * 144 Items
Product Details & Documents

LDPE, translucent, with white PP cap.

  • Good chemical resistance, heat resistant up to 80 °C
  • Bottle dispenses one drop at a time
  • Dropper tip snaps firmly in place and is easily removed for filling

With extended controlled dropper tip and dropper tip cap. These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications.

Specifications
  • Thread:
    20-410
  • Capacity:
    60 ml
  • Material:
    LDPE, white
  • Seq:
    08