Order Entry
Sweden
Orders LinkContactUsLinkComponent
Laboratory tables, without ground plate
Laboratory tables, without ground plate
Laboratory worktable, with left and right drawers, 1000×750×750 mm
Catalog # BOCH11500
Supplier:   BOCHEM
Laboratory tables, without ground plate
Catalog # BOCH11500
Supplier:  BOCHEM
Laboratory tables, without ground plate
Laboratory tables, without ground plate
Laboratory worktable, with left and right drawers, 1000×750×750 mm
Price
27 975,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
These laboratory tables are made up of 18/10 stainless steel and height adjustable.
  • Drawer face with user friendly handle bar
  • Noise protection
  • Worktops underlaid with soundproofing acoustic wood
Working plate with matt finishing and corners are continuously welded and polished.
Drawer with guard rails made of stainless steel to mount GN-drawer. Drawers can be taken off for cleaning, but are secured against drawing out.
Specifications
  • Description:
    Laboratory worktable, with left and right drawers
  • W×D×H:
    1000×750×750 mm