Order Entry
Sweden
Orders LinkContactUsLinkComponent
Dressing scissors
Dressing scissors
Blunt, 155 mm
Catalog # BOCH4108
Supplier:   BOCHEM
Dressing scissors
Catalog # BOCH4108
Supplier:  BOCHEM
Dressing scissors
Dressing scissors
Blunt, 155 mm
Price
84,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel. Multi-purpose scissors for a wide range of applications.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling
Specifications
  • Type:
    Blunt
  • Length:
    155 mm
  • Seq:
    09