Order Entry
Sweden
Orders LinkContactUsLinkComponent
Dressing scissors
Dressing scissors
Sharp, 145 mm
Catalog # 231-1293
Supplier:   BOCHEM
Dressing scissors
Catalog # 231-1293
Supplier:  BOCHEM
Dressing scissors
Dressing scissors
Sharp, 145 mm
Price
79,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel. Multi-purpose scissors for a wide range of applications.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Designed for rough handling
Specifications
  • Type:
    Sharp
  • Length:
    145 mm
  • Seq:
    05