0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SNABBVG OHAUS ADVENTURER PRO 810X0,01G 1 * 1 Items
Katalog # 611-0496
Leverantör:
OHAUS
Expand image
SNABBVG OHAUS ADVENTURER PRO 810X0,01G 1 * 1 Items
Katalog # 611-0496
Leverantör:
OHAUS