ChromBox kit
Deburring file
Catalog # JOURJR-802
Supplier: VICI JOUR RESEARCH
Jump to:
Product Details & Documents
The ChromBox is an all-purpose analytical spare parts kit for HPLC. It contains the most often used spare parts made both of high quality Type 316L SS as well as PEEK and PTFE.
: Kit includes PEEK, PTFE, and stainless steel tubing, PEEK and stainless steel nuts, ferrules and unions, tubing cutters and wrenches, all in a convenient carrying case.
Specifications
- Description:Deburring file
- Seq:04