Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TISSUE GRIND COMP SZ 19 1 * 1 items
Catalog # KIML885500-0019
Supplier:
KIMBLEKONTES
Expand image
TISSUE GRIND COMP SZ 19 1 * 1 items
Catalog # KIML885500-0019
Supplier:
KIMBLEKONTES