0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
TISSUE GRIND PESTLE SZ 21 1 * 1 items
KIML885451-0021
:
KIMBLEKONTES
TISSUE GRIND PESTLE SZ 21 1 * 1 items
KIML885451-0021
:
KIMBLEKONTES