0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GASTUBE DIMETHYL SULFIDE 0.25-10PPM 1 * 10 items
Catalog # GAST53
Supplier:
GASTEC
Expand image
GASTUBE DIMETHYL SULFIDE 0.25-10PPM 1 * 10 items
Catalog # GAST53
Supplier:
GASTEC