Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Y-27632, Millipore®

1 mg
Catalog # 688000-1
Y-27632, Millipore®
Catalog # 688000-1
CAS Number:  146986-50-7
Y-27632, Millipore®

1 mg

CAS Number:  146986-50-7
Price
3 025,00 kr
Log in for availability
1 * 1 mg
Product Details & Documents

A highly potent, cell-permeable, reversible, and selective inhibitor of Rho-associated protein kinases (Ki = 140 nm for p160ROCK). Also inhibits ROCK-II with equal potency. Also acts as a potent inhibitor of agonist-induced Ca2+ sensitization of myosin phosphorylation and smooth muscle contraction.

  • Inhibition is competitive with respect to ATP
  • Exhibits 10- to 50-fold lower affinity for PKCε than p160ROCK
  • Pretreatment with Y-27632 reduces the risk of lung injury in rat models of sepsis
  • Does not affect the activity of p21-activated protein kinase (PAK) even at higher concentrations (~100 µm)

Prevents apoptosis and enhances the survival and cloning efficiency of dissociated hES cells without affecting their pluripotency. A 5 mm (500 µg/296 µl) solution of Y-27632 in H2O and a 10 mm solution in DMSO are also available.

Synonyms:  trans-4-((R)-1-Aminoethyl)-N-(pyridin-4-yl)cyclohexanecarboxamide
  • CAS Number:146986-50-7
  • Formula:C₁₄H₂₁N₃O
  • MDL Number:MFCD03093855
  • Molecular Weight:247.34 g/mol
Specifications

Specifications

  • Pk:
    1 mg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.