Ion selective half-cell electrodes
4×10-7...1 mol/l, 2 - 9, 0...+50 °C, DX218-NH₄⁺, 2 mol/l MgSO₄, S7, NH₄⁺
Catalog # 662-2804
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
These ion selective half cells with solid-state membrane allow a quick and easy determination of concentration and activity of a variety of dissolved ions and gases.
- Modular and versatile ion selective half cells consisting of a universal shaft and a ion specific, exchangeable membrane
- Conform to RoHS (‘Restriction of Hazardous Substances’ in electrical and electronic equipment) and WEEE (‘Waste Electrical and Electronic Equipment’)
Ordering information: Membrane kits available separately.
Documents
Mettler InLab® Sensors
Product Literature
Specifications
- Concentration:4×10-7...1 mol/l
- Connection:S7
- For:NH₄⁺
- Type:DX218-NH₄⁺
- Electrolyte:2 mol/l MgSO₄
- Temp.:0...+50 °C
- pH:2 - 9
Accessories
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX280-Br⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX240-Ca²⁺
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX239-K⁺
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX262-NO₃⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX235-Cl⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX219-F⁻
Product Information
- Catalog #AvailabilityPrice
- Membrane kit for DX218-NH₄⁺
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



