Order Entry
Sweden
ContactUsLinkComponent
Thermometer, industrial, straight stem, mercury filled, measuring range:0:200/1C, capillary lens front, yellow backed, milk glass s cale, upper part:235x20mm, lower part: 105x8mm, cork-glass-top finish. 1 * 1 items
Katalog # 620-1614
Supplier:  Amarell
Thermometer, industrial, straight stem, mercury filled, measuring range:0:200/1C, capillary lens front, yellow backed, milk glass s cale, upper part:235x20mm, lower part: 105x8mm, cork-glass-top finish. 1 * 1 items
Katalog # 620-1614
Supplier:  Amarell