Order Entry
Sweden
ContactUsLinkComponent
Thermometer, to set Amarell universal, mercury filling, measuring range:147:202/0.1°C, dimension:400x9mm, enclosed scale, capillary lens front, total immersion, richter-top finish, suitable for government calibration. Until 10th October 2017 it is s 1 * 1 items
Catalog # 620-1062
Supplier:  Amarell
Thermometer, to set Amarell universal, mercury filling, measuring range:147:202/0.1°C, dimension:400x9mm, enclosed scale, capillary lens front, total immersion, richter-top finish, suitable for government calibration. Until 10th October 2017 it is s 1 * 1 items
Catalog # 620-1062
Supplier:  Amarell