0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Tubing, silicon, vacuum quality diameter(bottomx top) 8x16mm thick 4mm roll length 25m. 1 * 1 m
Catalog # 228-1578
Supplier:
DEUTSCH NEUMANN
Expand image
Tubing, silicon, vacuum quality diameter(bottomx top) 8x16mm thick 4mm roll length 25m. 1 * 1 m
Catalog # 228-1578
Supplier:
DEUTSCH NEUMANN