Order Entry
Sweden
ContactUsLinkComponent
TICKOPUR TR 7 - 25 LITRES, UNIVERSAL CLEANER, DEMULSIFYING, WITH CORROSION PROTECTION, CONCENTRATE, DOSAGE 0.1 - 5 %, MILDLY ALKALIN E, PH 8.9 (1 %) 1 * 25 L
Catalog # 142-1982
TICKOPUR TR 7 - 25 LITRES, UNIVERSAL CLEANER, DEMULSIFYING, WITH CORROSION PROTECTION, CONCENTRATE, DOSAGE 0.1 - 5 %, MILDLY ALKALIN E, PH 8.9 (1 %) 1 * 25 L
Catalog # 142-1982