Order Entry
Sweden
ContactUsLinkComponent
TICKOPUR R 33 - 25 LITRES, UNIVERSAL CLEANER, WITH CORROSION PROTECTION, CONCENTRATE, GENTLE TO MATERIAL, DOSAGE 1 - 5 %, MILDLY ALK ALINE, PH 9.9 (1 %) 1 * 25 L
Catalog # 142-1912
undefined
TICKOPUR R 33 - 25 LITRES, UNIVERSAL CLEANER, WITH CORROSION PROTECTION, CONCENTRATE, GENTLE TO MATERIAL, DOSAGE 1 - 5 %, MILDLY ALK ALINE, PH 9.9 (1 %) 1 * 25 L
Catalog # 142-1912