Order Entry
Sweden
ContactUsLinkComponent
Thermometer, mercury, ASTM1C, white backed, measuring range: -20:150/1C, length:322mm, 76mm immersion, durable pigment, government verified with verification certificate. Until 10th October 2017 it is still allowed to use mercury thermometers if th 1 * 1 items
Catalog # AMARA303011
Supplier:  Amarell
undefined
Thermometer, mercury, ASTM1C, white backed, measuring range: -20:150/1C, length:322mm, 76mm immersion, durable pigment, government verified with verification certificate. Until 10th October 2017 it is still allowed to use mercury thermometers if th 1 * 1 items
Catalog # AMARA303011
Supplier:  Amarell