Order Entry
Sweden
Orders LinkContactUsLinkComponent
Ferrules for gas chromatography
Ferrules for gas chromatography
Graphite/vespel ferrules, nut size 1/16", column i.Ø 0,32 mm, ferrule i.Ø 0,5 mm (2-Hole)
Catalog # PERCN9306000
Supplier:   PerkinElmer
Ferrules for gas chromatography
Catalog # PERCN9306000
Supplier:  PerkinElmer
Ferrules for gas chromatography
Graphite/vespel ferrules, nut size 1/16", column i.Ø 0,32 mm, ferrule i.Ø 0,5 mm (2-Hole)
Price
5 725,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Specifications
  • Description:
    Graphite/vespel ferrules, nut size 1/16", column i.Ø 0,32 mm, ferrule i.Ø 0,5 mm (2-Hole)