Order Entry
Sweden
Orders LinkContactUsLinkComponent
Ferrules for gas chromatography
Ferrules for gas chromatography
Graphite/vespel ferrules, nut size 1/8", internal Ø 1/8"
Catalog # PERCN9302081
Supplier:   PerkinElmer
Ferrules for gas chromatography
Catalog # PERCN9302081
Supplier:  PerkinElmer
Ferrules for gas chromatography
Graphite/vespel ferrules, nut size 1/8", internal Ø 1/8"
Price
3 425,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Specifications
  • Description:
    Graphite/vespel ferrules, nut size 1/8", internal Ø 1/8"